Gibco™

Essential 8™ Medium

Catalog number:  A1517001

 Related applications: Clinical & Translational Research | Stem Cell Culture | Stem Cell Engineering & Reprogramming

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Essential 8™ Medium is a xeno-free and feeder-free medium specially formulated for the growth and expansion of human pluripotent stem cells (PSCs). Originally developed by Guokai Chen et al. in the laboratory of James Thomson (published as "E8") and validated by Cellular Dynamics International, Essential 8™ Medium has been extensively tested and proved to maintain pluripotency in multiple iPSC lines. In addition, Essential 8™ Medium has been used to scale up production of iPSCs and shown to support iPSC growth for >50 passages without any signs of karyotypic abnormalities, along with maintaining the ability of iPSCs to differentiate into all three germ line lineages.

Features of Essential 8™ Medium include:

• Consistent—reduced variability compared to existing feeder-free culture media
• Robust—reliable and robust cultures with a xeno-free, cGMP, 8-component medium
• Cost effective—economical and scalable PSC culture compared to other feeder-free media

Reduced Variability
Essential 8™ Medium is xeno-free and contains only the 8 essential components needed for stem cell culture. Unlike other media that contain over 20 highly variable ingredients, Essential 8™ Medium is produced under cGMP and has an optimized formulation and growth factor levels to help ensure maximum cell health, pluripotency, and growth, with minimal variability.

Reliable and Robust Cultures
Essential 8™ Medium has been show to support pluripotent stem cell growth and provide cultures with superior morphology and growth kinetics compared to other feeder systems.

Cost Effective
Essential 8™ Medium is provided in a convenient two-component kit (500 ml basal medium & 10 ml supplement), and when used with vitronectin (VTN-N), provides a cost effective, defined system for feeder-free culture of human pluripotent stem cells (PSCs).

Essential 8™ Medium is commercialized in partnership with Cellular Dynamics International.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Manufacturing Quality: cGMP for medical devices, 21 CFR Part 820 and ISO 13485
Cell Type: Stem Cells (Embryonic), Stem Cells (Human), Stem Cells (iPS - Induced Pluripotent Stem)
Classification: Xeno-Free
Culture Type: Feeder Cell-Free
Form: Liquid
Phenol Red Indicator: Phenol Red
Product Line: Essential 8™
Product Size: 500 mL
Quantity: 1 each
Serum Level: Serum-Free
Species: Human
Volume (Metric): 500 ml

Contents & storage

500 mL basal medium (store at 2—8°C)
10 mL supplement (store at -5 to -30°C)

Documents

Manuals & protocols

Help center

Cell Culture & Transfection Learning Center

Access cell culture and transfection educational resources for better experiment planning and execution.

Cell Culture & Transfection Support Centers

Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.

Recommended products