StemPro® Neural Stem Cells

Catalog number: A15654

Gibco™  Related applications: Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

StemPro® Neural Stem Cells are cryopreserved human fetal brain-derived neural stem cells (NSCs). These cells are manufactured under Good Manufacturing Practice (GMP) manufacturing standards in a California-licensed facility and are available for Research Use Only. They are isolated and expanded in culture to passage 4 under a low oxygen (ischemic tolerant, proprietary manufacturing process using FBS-supplemented media and then cryopreserved in an animal origin-free cryopreservation medium. The cells are supplied at a concentration of 1 x 106 viable cells/vial. A Certificate of Analysis is available for each lot that documents the quality control specifications, test results, and donor information. These cells have no observed tumorigenicity/toxicity in GLP-compliant animal studies.

StemPro® Neural Stem Cells help you to:

• Minimize assay variability with itNSCs manufactured using GMP manufacturing standards
• Improve functionality studies with immature, high potency NSCs
• Complete experiments requiring one large lot of NSCs
• Minimize your chance of assay failure by using tested cells, reagents, and protocols

Immature, High Potency, Low Oxygen-Cultured NSCs
StemPro® Neural Stem Cells are derived from human fetal brain from qualified, traceable donors. The cells are isolated, cultured, and expanded under Good Manufacturing Practice (GMP) manufacturing standards in a California-licensed facility using a proprietary Reduced Oxygen Tension manufacturing process. Manufacture of cells in a reduced oxygen tension environment results in higher yields of highly potent immature stem cells compared to cells expanded in normal oxygen culture conditions.

Safety Tested
StemPro® Neural Stem Cells have been demonstrated to be non-toxic and non-tumorigenic in GLP animal studies performed according to FDA guidelines.

Large lots can be made available for those requiring assay standardization. Protocols using tested reagents are provided to help minimize your chance of assay failure using reagents and cells that have not been evaluated to work together. View our tested media and reagents.

High Post-Cryopreservation Viability
These StemPro® Neural Stem Cells are supplied in CryoStor® cryopreservation medium at a concentration of 1x106 viable cells/vial. CryoStor® medium is animal origin-free and shown to support high cell viabilities post-thawing. Controlled processes are used for cryopreservation, handling, and shipping to help minimize risk of adverse effects to the quality of the cells.

Made in USA by Stemedica for Thermo Fisher Scientific.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Age: Fetal
Form: Cryopreserved
Species: Human
Quantity: 1 vial(s)
Cell Type: Stem Cells (Neural)
Culture Type: Spheroid Culture
Product Size: 1 x 106 cells
Classification: FBS Supplemented
Parental Cells: Primary Tissue
Number of Cells: 1 x 106
Donor Attributes: Normal
Cell Passage Number: Passage 4
Media Recommendation: StemPro® NSC SFM

Contents & storage

1 tube. Store in liquid nitrogen.

Documents

Manuals & protocols