PolarScreen™ ER Beta Competitor Assay Kit, Red

Catalog number: A15891

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PolarScreen™ ER Beta Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential estrogen receptor (ER) beta ligands. The kit uses insect cell-expressed, full-length, native (untagged) human ER beta (A15664) and a novel, high-affinity, fluorescent estrogen ligand (Fluormone™ ES2) in a homogenous, mix-and-read assay format.

This kit produces a red color read-out; a "green" kit is also available (A15890). Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance email drugdiscoverytech@lifetech.com.

The PolarScreen™ ER Beta Competitor Assay Kit is:

• Complete & ready-to-use out of the box—all reagents included, just add your test compounds
• High throughput compatible—20 µL fluorescence polarization assays in a 384-well plate
• Convenient—available in bulk sizes for larger screening needs

How the Assay Works
Full-length ER beta is added to a fluorescent estrogen ligand to form an ER-Fluormone™ ES2 complex. This complex is added to individual test compounds in multiwell plates. To determine relative affinity, the assay measures fluorescence polarization. Competing test compounds will displace the Fluormone™ ES2 ligand from ER beta, permitting it to tumble rapidly and resulting in low polarization values. A shift in the fluorescence polarization value in the presence of test compound is used to determine the relative affinity of test compounds for ER beta.

All components are manufactured under the strictest quality parameters. This product's performance, per the Certificate of Analysis, is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Red
Druggable Target: Nuclear Receptors
Format: 384-well plate
Product Line: PolarScreen™
Product Size: 1 kit
Readout: Fluorescence Polarization
Target Gene: Estrogen Receptor Beta Nuclear Receptor
Technique: Fluorescence Polarization
Validated Application: Nuclear Receptor Assay

Contents & storage

• Estrogen Receptor (ER) Beta Recombinant Protein, Full Length (store at -68 to -85°C)
• Fluromone™ EL Red (store at -5 to -30° C)
• ER Red Assay Buffer (store at room temperature)

Documents

Manuals & protocols