LanthaScreen® TR-FRET ER beta Competitive Binding Kit
Catalog number: A15895
 Related applications: Target & Lead Identification & Validation
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The LanthaScreen® TR-FRET ER beta Competitive Binding Kit provides a sensitive and robust method for high-throughput screening (HTS) of ligands for estrogen receptor (ER) beta. The kit uses a human ER beta ligand-binding domain (LBD) that is tagged with glutathione S-transferase (GST), a terbium-labeled anti-GST antibody, and a fluorescent small-molecule ER ligand (Fluormone™ ES2) in a homogeneous mix-and-read assay format.
The Lanthascreen ER beta Competitive Binding Assay is:
• Complete & ready-to-use out of the box—all reagents included, just add your test compounds
• High throughput compatible—TR-FRET assays in a 384-well plate
• Convenient—components available in bulk sizes for larger screening needs
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Size: 800 x 20 µL assays
Validated Application: Competitive Binding Assay, Nuclear Receptor Assay
Contents & storage
1 tube ER beta Ligand Binding Domain Recombinant Protein (store at -68 to -85°C)
1 tube of Fluormone ES2 (store at -5 to -30°C)
2 bottles NR Buffer (store at -5 to -30°C)
1 tube Tb-Anti-GST Ab (store at -5 to -30°C)
1 tube of DTT (store at -5 to -30°C)
Documents
Manuals & protocols