PolarScreen™ Vitamin D Receptor Competitor Assay Kit, Red

Catalog number:  A15907

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PolarScreen™ Vitamin D Receptor Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential vitamin D receptor (VDR) ligands. The kit uses insect cell-expressed, human full-length VDR (also available separately) and a novel, high-affinity, fluorescent VDR ligand (Fluormone™ VDR Red) in a homogenous, mix-and-read assay format.

This kit produces a red color read-out and is suitable to determine the IC50s of competitive compounds. For additional information or assistance contact us at drugdiscoverytech@lifetech.com.

The PolarScreen Vitamine D Receptor Competitor Assay is:

• Complete & ready-to-use out of the box—all reagents included, just add your test compounds
• High throughput compatible—fluorescence polarization assays in a 384-well plate
• Convenient—components available in bulk sizes for larger screening needs

How the Assay Works
VDR is added to a fluorescent VDR ligand (Fluormone VDR Red) to form a receptor/tracer complex, resulting in a high polarization value. This complex is then added to individual test compounds in microwell plates. Competitors displace the tracer from the complex, causing the fluorescent ligand to tumble more rapidly during its fluorescence lifetime, resulting in a low polarization value. Noncompetitors will not displace the fluorescent ligand from the complex; therefore, the polarization value will remain high. The shift in polarization value in the presence of test compounds is used to determine relative affinity of test compounds for VDR (see figure).

Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: PolarScreen™
Product Size: 800 x 20 µL assays
Validated Application: Competitive Binding Assay, Nuclear Receptor Assay

Contents & storage

1 tube VDR Recombinant Human Protein (store at -68 to -85°C)
1 tube of Fluormone VDR Red Tracer (store at -5 to -30°C)
3 bottles of VDR Red Screening Buffer (store at -5 to -30°C)
1 tube of DTT (store at -5 to -30°C)

Documents

Manuals & protocols