CD28 Antibody (37.51), APC conjugate, Molecular Probes™

Catalog number: A16222

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. It is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes (approximately 95% of CD4+ cells and 50% of CD8+ cells). Mature thymocytes exhibit higher levels of CD28 than the immature cells. Activation of T cells results in enhanced CD28 expression. T cell activation requires two combined signals provided by antigen presenting cells.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Mouse
Validated Application: Flow Cytometry
Host Species: Hamster
SubClass (Isotype): IgG
Clonality: Monoclonal
Clone ID: 37.51
Antibody Fragment: Whole Antibody
Target Gene: Cd28
Gene ID: 12487
Gene Aliases: CH29-189K17.3
Label or Dye: APC (Allophycocyanin)
Excitation⁄Emission (nm): 650⁄660
Excitation Class: Visible
Concentration: 0.2 mg⁄ml
Form: Liquid
Number of Reactions: 100 Tests
For Use With (Equipment): Attune® Acoustic Focusing Cytometer
Product Size: 50 µg
Other Additives: Sodium Azide Preservative

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products