CD90.1 / Thy-1.1 Antibody (53-2.1), PE-Cy5® conjugate, Molecular Probes™

Catalog number: A16408

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. It is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain.

Specifications

Product Size: Case of 960
认证/合规: Manufactured according to ISO 9001, ISO 14001 and ISO 13485
颜色: Violet
描述: ClipTip 50, nonsterile
带过滤器: No
平台长度(英制): 2.01 in.
长度(公制): 5.1cm
包装: 10 x 96/rack
Sterility: Nonsterile
容积(公制): 50μL
适用于(设备): Thermo Scientific F1-ClipTip manual pipette, E1-ClipTip electronic pipette
Product Line: ClipTip

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products