Invitrogen™

Alignflow™ Flow Cytometry Alignment Beads for Blue Lasers, 2.5 μm

Catalog number: A16500
Invitrogen™

Alignflow™ Flow Cytometry Alignment Beads for Blue Lasers, 2.5 μm

Catalog number: A16500
Catalog Number
A16500
also known as A-16500
Unit Size
3 mL
Price (USD)
Price: 396.00
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A16500
also known as A-16500
3 mL
Price: 396.00
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Molecular Probes™ AlignFlow™ flow cytometry alignment beads are reliable references for aligning, focusing, and calibrating flow cytometers.The 2.5 μm Alignflow™ Flow Cytometry Alignment Beads for Blue Lasers are fluorescent-dye infused microspheres that are excitable at 488 nm with emission in the range of 515–660 nm.

View information for all flow cytometry calibration and size reference beads.

AlignFlow™ beads provide:

• Uniformity—very low FSC CV and intensity CV
• Consistency in production, creating ideal reference standards
• Confidence through reliable, optimal instrument performance
• Compatibility—use with any instrument

The AlignFlow™ beads are highly uniform with respect to size and fluorescence intensity, resulting in low FSC CV and intensity CV. The beads approximate the size, emission wavelength, and intensity of many biological samples and permit the calibration of the flow cytometer’s laser source, optics, and stream flow without wasting valuable and sensitive experimental material.

Bead Labeling
As with all of our flow cytometry standard microspheres, the AlignFlow™ beads are stained internally rather than on the surface. The dyes are therefore insulated from environmental interactions that could cause variable fluorescence output, resulting in excellent signal stability. These fluorescent polystyrene microspheres are supplied as suspensions packaged in dropper vials for convenient dispensation and are available in a choice of three colors (UV, blue, red) and two sizes (2.5 μm, 6.0 μm) to match your laser.

Notes:
The AlignFlow™ flow cytometry reference beads are provided in updated packaging that includes a Quick Reference Label that puts key information for product use at your fingertips.

These beads replace previous AlignFlow™ flow cytometry reference bead products. This particular product replaces AlignFlow™ Flow Cytometry Alignment Beads, 2.5 μm, for 488 excitation (A7302).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescent
Diameter (Metric)
2.5 μm
For Use With (Equipment)
Flow Cytometer
Format
Suspension
Shipping Condition
Room Temperature
Stain Configuration
Core
Product Line
AlignFlow™
Product Type
Alignment Beads

Contents & Storage

Contains 2 vials of Alignflow™ beads (1.5 mL per vial). Store at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text