Ly6G / GR-1 Antibody, PE-Cy5.5® conjugate (RB6-8C5)

Catalog number: A18508

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Specifications

Reactivity: Mouse
Validated Application: Flow Cytometry
Host Species: Rat
SubClass (Isotype): IgG2b, kappa
Clonality: Monoclonal
Clone ID: RB6-8C5
Antibody Fragment: Whole Antibody
Target Gene: lymphocyte antigen 6 complex, locus G
Gene ID: 546644
Gene Symbol: Ly6g
Gene Aliases: Gr-1, Gr1, Ly-6G
Label or Dye: PE-Cy®5.5
Excitation⁄Emission (nm): 488⁄694
Excitation Class: Visible
Concentration: 0.2 mg⁄ml
Form: Liquid
Number of Reactions: 800 Tests
For Use With (Equipment): Attune® Acoustic Focusing Cytometer
Product Size: 100 µg
Other Additives: Sodium Azide Preservative

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products