Oncomine™ Concepts Edition, Biotech License
Catalog number:  A18566
 Related applications: Cancer Genomics
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Oncomine™ Concepts Edition is a powerful web application for biotech and pharma scientists that integrates and unifies high-throughput cancer profiling data so that target expression across a large volume of cancer types, subtypes, and experiments can be assessed online, in seconds. Oncomine™ Concepts Edition is a hypothesis-neutral, data driven solution developed to leverage datasets from global sources and consists of the most comprehensive collection of oncology focused and curated datasets.
Oncomine™ Concepts Edition is complete with analysis and visualization methods designed to help scientists address the most relevant drug discovery and biomarker discovery questions. It includes thousands of pre-computed analyses for differential expression, co-expression, outlier analysis, as well as meta-analyses to compare independent results on differential expression and outliers. Additionally, it allows users to search the Oncomine™ database for enrichment of user-defined gene signatures. These analyses provide valuable insight into biology, regulation, pathways, drug response, and patient populations. Overall, Oncomine™ Concepts Edition provides a simple yet powerful way to support target discovery, validation, and the prioritization of tumor populations for drug development.
Learn more about Oncomine™ Concepts Edition.
Oncomine™ Concepts Edition can be licensed annually for 10 users or for a global, unlimited number of users. Oncomine™ Concepts Edition requires the Oncomine™ Data Bundle #1 license.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
License Type: Annual, Commercial/Pharma, Multi-Users
Product Line: Oncomine™
Product Size: 1 year, 10 users
Software Category: Software
Documents
Manuals & protocols