New Genome Editing Support Center—Find technical tips and troubleshooting recommendations from our scientists.
Invitrogen™

GeneArt™ CRISPR Nuclease Vector with CD4 Enrichment Kit (with competent cells)

Catalog number:  A21177

 Related applications: Seamless Cloning & Genetic Assembly

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The GeneArt® CRISPR Nuclease Vector with CD4 Enrichment Kit is a vector system for expression of the functional components needed for CRISPR/Cas9 genome editing in mammalian cells with a CD4 reporter. The CD4 reporter enables bead-based enrichment, an option for magnetic bead-based sorting/enrichment of Cas9 & CRISPR expressing cells using Dynabeads® CD4 magnetic beads. Transfection efficiency can also be tracked using anti-CD4 fluorescent antibodies. The linearized GeneArt® CRISPR nuclease vectors provide a rapid and efficient way to clone double-stranded oligonucleotides encoding a crRNA representing a desired target into an expression cassette that allows sequence-specific targeting of the Cas9 nuclease. This version of the kit includes OneShot® TOP10 Competent Cells. A version without competent cells is also available (Cat. No. A21175).

The GeneArt® CRISPR Nuclease Vector system with competent cells offers:

• Easy-to-design genome engineering system
• Affordable, ready-to-use cloning vectors
• Enrichment for hard to transfect or difficult to modify cell lines
• Competent cells included for added convenience and highest cloning efficiencies

All-in-One Vector System for CRISPR-based Genome Editing
The GeneArt® CRISPR Nuclease Vector kit offers a simple, ready-to-use, all-in-one expression vector system consisting of both a Cas9 nuclease expression cassette and a guide RNA (gRNA) cloning cassette for rapid and efficient cloning of DNA that encodes target-specific CRISPR RNA (crRNA). This system allows you to edit and engineer a genomic locus of choice in a sequence-specific manner from a single plasmid. After relevant targets have been identified with fast and easy-to-use GeneArt® CRISPR vectors, the biologically relevant mutations can be precisely created with GeneArt® Precision TALs, with high specificity and low off-target effects.

Need assistance with CRISPR gRNA design?
Our new CRISPR Search & Design tool allows you to search our database of >600,000 predesigned CRISPR gRNAs in human and mouse genes or analyze your sequence of interest for de novo gRNA designs using our proprietary algorithms. CRISPR sequences are optimized for gene knockout and target the first three transcribed exons for each gene. Search results include the top 6 CRISPR sequences with PAM sites, exon maps with gRNA binding sites, and potential off-target binding sites for each CRISPR sequence. The tool will design the correct gRNA format for your preferred CRISPR-Cas9 product, including oligos for your GeneArt® CRISPR Nuclease Vector. Start designing today >

Resources
Demonstrated protocol: Engineering stem cells with CRISPR-Cas9
Demonstrated protocol: CRISPR-Cas9 microinjection in mice and zebrafish embryos

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cloning Method: Restriction Enzyme ⁄ MCS
Delivery Method: Transfection
Key Function: Episomal Expression, Gene Editing, Genome Engineering, Stable Cell Line Development
Product Line: GeneArt®
Product Size: 10 reactions
Promoter: CMV, U6
Protein Tag or Fusion: CD4 fusion
Reporter: CD4

Contents & storage

• CRISPR CD4 Nuclease Vector, linearized
• 10X Oligonucleotide Annealing Buffer
• DNase/RNase-free Water
• 5X Ligation Buffer
• T4 DNA Ligase
• U6 Forward Sequencing Primer
• Control, double stranded oligonucleotide
Store the above at -5 to -30°C.

• OneShot® TOP10 Chemically Competent Cells
Store at -80°C.

Documents

Manuals & protocols