See more signal from immunofluorescence images with Prolong® Diamond. Learn more.

Goat anti-Rabbit IgG (H+L) Secondary Antibody, Alexa Fluor® 647 conjugate

Catalog number: A21244

Invitrogen™  Related applications: Secondary Detection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Rabbit IgG (H+L) Secondary Antibody for IF, Flow
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Labeling Method: Zenon® Secondary Detection-Based
Color: Far-Red
Reactivity: Mouse
Label or Dye: Alexa Fluor™ 647
Product Size: 1 kit
Emission Class: near IR
Labeling Scale: < 1-20 µg
Target Isotype: IgG2b
Labeling Target: Antibodies (General)
Detection Method: Fluorescent
Shipping Condition: Room Temperature
Excitation⁄Emission (nm): 650⁄665

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products