Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
BioProbes™ Journal of Cell Biology Applications—Subscribe today »
Invitrogen™

Amplex™ Red Hydrogen Peroxide/Peroxidase Assay Kit

Catalog number:  A22188

 Related applications: Cell Viability, Proliferation & Function | Protein Assays and Analysis | Stem Cell Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Amplex® Red Hydrogen Peroxide/Peroxidase Assay Kit contains a sensitive, one-step assay that uses the Amplex® Red reagent (10-acetyl-3,7-dihydroxyphenoxazine) to detect hydrogen peroxide (H2O2) or peroxidase activity. The Amplex® Red reagent, in combination with horseradish peroxidase (HRP), has been used to detect H2O2 released from biological samples, including cells, or generated in enzyme-coupled reactions. Furthermore, Amplex® Red reagent can be used as an ultrasensitive assay for peroxidase activity when H2O2 is in excess. In the presence of peroxidase, the Amplex® Red reagent reacts with H2O2 in a 1:1 stoichiometry to produce the red-fluorescent oxidation product, resorufin. Resorufin has excitation and emission maxima of approximately 571 nm and 585 nm, respectively, and because the extinction coefficient is high (58,000 ± 5,000 cm–1M–1), you can perform the assay fluorometrically or spectrophotometrically. This reaction has been used to detect as little as 10 picomoles of H2O2 in a 100 µL volume or 1 x 10–5 U/mL of HRP.

Amplex® Red Hydrogen Peroxide/Peroxidase Assay Kit Specifications:
• Ex/Em of reaction product resorufin: ~571/585 nm
• Sensitive one-step assay to detect hydrogen peroxide or peroxidase in vitro
• Can detect as little as 10 picomoles of hydrogen peroxide or 10 µU/mL of horseradish peroxidase activity in a 100 µL assay volume
• Provides sufficient reagents for approximately 500 assays using reaction volumes of 100 µL per assay
• Designed for a 96-well fluorescence or absorbance microplate reader, but can be easily adapted for other volumes, for fluorimeters, and for spectrophotometers


Find More Substrates for Oxidases
Review Substrates for Oxidases, Including Amplex Red Kits—Section 10.5 in the Molecular Probes® Handbook for more information on these products.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Fluorometer, Microplate Reader, Spectrophotometer
Detection Method: Fluorescent
Format: 96-well plate, Tube(s)
Label or Dye: Amplex® Red
Product Line: Amplex™
Product Size: 500 assays
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols