Get up to 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Alexa Fluor™ 633 Phalloidin

Catalog number:  A22284

Invitrogen™  Related applications: Cell Structure

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Alexa Fluor® 633 phalloidin is a high-affinity F-actin probe conjugated to our bright, photostable, far-red fluorescent Alexa Fluor® 633 dye.

Selectively stains F-actin
Outstanding fluorescence performance
Excitation/Emission: 632/647 nm
Superior to antibody staining
Optimal for fixed and permeabilized samples

Get Superior Results in Actin Staining Studies
Phalloidin is a bicyclic peptide belonging to a family of toxins isolated from the deadly Amanita phalloides "death cap" mushroom and is commonly used in imaging applications to selectively label F-actin. Fluorescently-labeled phalloidin has virtually identical binding properties with actin from different species including plants and animals. Phalloidin binds F-actin with high selectivity while Alexa Fluor® 633 provides far-red fluorescence of unparalleled brightness and photostability. Demonstrating very little nonspecific staining, Alexa Fluor® 633 phalloidin allows high-contrast discrimination of actin staining.

Use in Multiple Applications
Alexa Fluor® 633 phalloidin can be used to visualize and quantitate F-actin in tissue sections, cell cultures, or cell-free preparations. Alexa Fluor® 633 phalloidin staining is fully compatible with other fluorescent stains used in cellular analyses including fluorescent proteins, Qdot® nanocrystals, and other Alexa Fluor® conjugates including secondary antibodies.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Links
Read about labeling F-Actin with phallotoxins.
See the full line of Alexa Fluor® Dye products.
Create a Cellular Masterpiece with Alexa Fluor® Phalloidin using the Cell Paint Tool.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

LoyaltyEligible: SKUEligibleForLoyalty
Label or Dye: Alexa Fluor® 633
Sub-Cellular Localization: Actin, Cytoskeleton
Excitation⁄Emission (nm): 632⁄647
Product Line: Alexa Fluor™
Product Size: 300 units
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products