MAX Efficiency™ Transformation Reagent for Algae

Catalog number:  A24229

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

MAX Efficiency® Transformation Reagent for Algae is a buffer that enhances transformation efficiency for multiple strains of Chlamydomonas species. One of the biggest hurdles in research and development with Chlamydomonas has been the introduction of exogenous DNA into Chlamydomonas strains due to rigid cell walls. Methods such as glass bead agitation, electroporation, and microparticle bombardment are available but often result in low transformation efficiency.

MAX Efficiency® Transformation Reagent for Algae increases the permeability of the Chlamydomonas cell wall and facilitates increased delivery of DNA into the cell nucleus by electroporation. Simply grow the Chlamydomonas cells to early log phase, harvest by centrifugation, and wash twice with MAX Efficiency® Transformation Reagent prior to electroporation. To date, we have seen >200-fold increase in transformation efficiency over previously recommended conditions in 10 different Chlamydomonas strains, including wild type and mutants, using circular or linear DNA, as well as PCR fragments.

• Obtain >1000 transformants/µg DNA for cell wall(+) strains
• Obtain >100 transformants/µg DNA for cell wall(-) strains
• Transform circular or linear DNA or PCR fragments
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line: Chlamydomonas
Detection Method: Gel Electrophoresis
Expression: Algae
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Number of Reactions: 40 Reactions
Product Line: GeneArt®
Product Size: 40 reactions
Selection Agent (Eukaryotic): Hygromycin

Contents & storage

Contains:
1 bottle Transformation Reagent, 250 mL

Documents

Manuals & protocols