Foxp3 Transcription Factor Wash Buffer

Catalog number: A24261

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Foxp3 Transcription Factor Wash Buffer has been specially formulated to work with the APC (A18629), FITC (A18662), and PE (A18690) conjugates of Foxp3 Mouse Anti-Mouse Monoclonal Antibody (clone 3G3). This buffer is used during the wash and antibody staining steps following fixation and permeabilization with a 1X solution of Foxp3 Transcription Factor Fixation and Permeabilization Buffer A (A24217, a 4X concentrate) and Foxp3 Transcription Factor Fixation and Permeabilization Buffer B (A24218, a diluent), for optimal intranuclear staining with Foxp3-conjugated antibodies used in flow cytometry applications.

How It Is Used
The Foxp3 Transcription Factor Wash Buffer is supplied as a 10X stock solution. The 10X stock solution should be diluted to a 1X working concentration in PBS containing 1% BSA and used for wash and Foxp3 staining steps following fixation and permeabilization with 1X Foxp3 Fixation and Permabilization Buffer (1 part Buffer A: 3 parts Buffer B).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Molecular Probes®
Product Size: 100 mL
Reagent Type: Fixation Reagent, Permeabilization Reagent

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products