New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

Human Neural Stem Cell Immunocytochemistry Kit

Catalog number:  A24354

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Molecular Probes® Human Neural Stem Cell Immunocytochemistry Kit enables convenient image-based analysis of four common markers of human neural stem cells (NSCs): nestin, PAX6, SOX1, and SOX2. This complete immunocytochemistry (ICC) kit includes an optimized set of primary and secondary antibodies, a nuclear DNA stain, and all of the buffers necessary to complete the staining protocol.

Easily scalable to any culture plate, dish, or slide format, the Human Neural Stem Cell Immunocytochemistry Kit comes with reagents sufficient to run 20 samples using a 50 µL staining volume.

• Confirm expression of key neural stem cell markers with highly specific antibodies
• Get more information per sample by measuring more than one marker at a time
• Generate beautiful multiplexed images with minimal effort
• Process samples with confidence using a complete & optimized set of ICC reagents

Confirm Expression of NSC Markers
The antibodies included in the kit have been carefully selected for high performance in ICC analysis of NSCs. These antibodies are well suited for the characterization of NSCs generated using Gibco® PSC Neural Induction Medium.

Get More Information Per Sample
Save time and precious sample. The antibody combinations in this kit have been designed to allow for simultaneous assessment of two markers at a time (e.g., nestin & SOX2 or SOX1 & PAX6) along with nuclear DNA staining.

Generate Beautiful Multiplexed Images
Powered by the superior performance of bright, photostable Alexa Fluor® dyes, this kit enables the acquisition of beautiful multiplexed images of NSC marker expression using commonly available blue, green, and orange/red filter sets.

Process Samples with Confidence
Pre-tested fixation, permeabilization, blocking, and wash buffers allow you to focus on answering scientific questions rather than spending time determining reagent compatibility.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Alexa Fluor™ 488, Alexa Fluor™ 555, Alexa Fluor™ 594
Product Size: 20 tests

Contents & storage

Contains:
• 20 µL of Goat Anti-Human SOX1 (50X)
• 20 µL of Rabbit Anti-Human SOX2 (50X)
• 20 µL of Mouse Anti-Human Nestin (50X)
• 20 µL of Rabbit Anti-Human PAX6 (50X)
• 20 µL of Alexa Fluor® 488 Anti-Mouse (250X)
• 20 µL of Alexa Fluor® 488 Anti-Goat (250X)
• 40 µL of Alexa Fluor® 555 Anti-Rabbit (250X)
• 40 µL of Alexa Fluor® 594 Anti-Rabbit (250X)
• 1 vial of NucBlue® Fixed Cell Stain ReadyProbes™ Reagent (1X)
• 10 mL of Fixative Solution (1X)
• 10 mL of Permeabilization Solution (1X)
• 20 mL of Blocking Solution (1X)
• 20 mL of Wash Buffer (10X)

Documents

Manuals & protocols

Recommended products