Applied Biosystems™

JUN™ Dye Spectral Calibration Plate for Multiplex qPCR, 384-well

Catalog number:  A24733

 Related applications: Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This plate is used for JUN® dye spectral calibration of your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, and QuantStudio™ 12K Flex real-time PCR systems with a 384-well block. Calibration and verification should be run at least every six months. Please see your instrument maintenance guide for full instructions on use. The dye formulations will improve results with multiplexing by more accurately representing fluorescent spectra found in your real-time PCR experiments.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Calibration Type: Spectral Calibration
For Use With (Equipment): QuantStudio™, ViiA™ 7 System
Label or Dye: JUN™
Product Size: 1 plate

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products