TRA-1-60 Alexa Fluor® 488 Conjugate Kit for Live Cell Imaging

Catalog number: A25618

Invitrogen™  Related applications: Cellular Imaging | Stem Cell Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Molecular Probes® human pluripotent stem cell (PSC) live imaging kits provide reagents for superior imaging of live human PSCs. This TRA-1-60 Alexa Fluor® 488 Conjugate Kit includes an optimally labeled primary antibody paired with reduced-background imaging medium (FluoroBrite™ DMEM) for the specific detection of the PSC marker TRA-1-60. For fixed cell staining of PSC markers and germ layer markers, we recommend the Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit.

With the TRA-1-60 Alexa Fluor® 488 Conjugate Kit for Live Cell Imaging, you can:
• Confirm the expression of a key pluripotent stem cell marker using an optimally dye-conjugated antibody
• Preserve cell health and maximize fluorescence signal with FluoroBrite™ DMEM
• Stain live cell samples with confidence using reagents that are tested to be free of common contaminants

Confirm Expression of a Key PSC Marker
Powered by the high performance of bright, photostable Alexa Fluor® dyes, this kit enables reliable live-cell detection of the surface PSC marker TRA-1-60. This antibody conjugate has been demonstrated to show superior staining results when used with FluoroBrite™ DMEM.

Preserve Cell Health and Maximize Signal
Staining cells in FluoroBrite™ DMEM maximizes cell health and enhances signal-to-noise ratio when performing live-cell fluorescence imaging.

Stain Live-cell Samples with Confidence
Unlike most other conjugated antibodies, the reagents provided in this kit are 0.2 µm sterile-filtered and tested to be free of common contaminants.

What You Get
Easily scalable to any culture plate, dish, or slide format, the TRA-1-60 Alexa Fluor® 488 Conjugate Kit comes with reagents sufficient to stain 50 samples using a 200 µL staining volume and includes:
• 200 µL of Alexa Fluor® 488 Mouse Anti-Human TRA-1-60 (50X concentration)
• 500 mL FluoroBrite™ DMEM
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Alexa Fluor™ 488
Product Line: Alexa Fluor®
Product Size: 50 tests

Contents & storage

• 200 µL of Alexa Fluor® 488 Mouse Anti-Human TRA-1-60 (50x concentration) (store at 2–8°C)
• 500 mL FluoroBrite™ DMEM (store at 2–8°C, protect from light)

Documents

Manuals & protocols

Recommended products