Mini Gel Tank Lid

Catalog number: A25944

Mini Gel Tank Lid

Catalog number: A25944
Catalog Number
A25944
Unit Size
1 unit
Price (USD)
Price: 198.00
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A259441 unit
Price: 198.00
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Mini Gel Tank Lid is a replacement part for the Mini Gel Tank. It goes on top of the Gel Runner Tank.

About the Mini Gel Tank
The Mini Gel Tank is compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Mini Gels
Gel Compatibility
Novex™ Mini Gels
Gel Size
Mini
Type
Mini Gel Tank Lid
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text