Invitrogen™

Alexa Fluor™ Plus 405 Phalloidin

Catalog number: A30104
Invitrogen™

Alexa Fluor™ Plus 405 Phalloidin

Catalog number: A30104
Catalog Number
A30104
Unit Size
1 vial
Price (USD)
Price: 636.00
Online price: 622.65
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A301041 vial
Price: 636.00
Online price: 622.65
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Alexa Fluor Plus 405 Phalloidin is a high-affinity filamentous actin (F-actin) probe (phalloidin) conjugated to our bright, photostable, violet-fluorescent Alexa Fluor 405 dye.

• Selectively stains F-actin
• Outstanding fluorescence performance
• Excitation/emission: 405/450 nm. Compatible with standard DAPI filter set or 405/violet excitation laser.
• Superior to antibody staining
• Optimal for fixed and permeabilized samples

Get superior results in your actin staining studies
Phalloidin is a bi-cyclic peptide belonging to a family of toxins isolated from the deadly Amanita phalloides ''death cap'' mushroom and commonly used in imaging applications to selectively label F-actin. Fluorescently-labeled phalloidin has several advantages over antibodies for actin labeling, including virtually identical binding properties with actin from different species of plants and animals, and low non-specific binding. Phalloidin binds F-actin with high selectivity, while Alexa Fluor Plus 405 provides violet fluorescence of unparalleled brightness and photostability.

Use in multiple applications
Alexa Fluor Plus 405 Phalloidin can be used to visualize and quantitate F-actin in tissue sections, cell cultures, or cell-free preparations. Its staining is fully compatible with other fluorescent stains used in cellular analyses, including fluorescent proteins, Qdot nanocrystals, and other Alexa Fluor conjugates including secondary antibodies.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Excitation Wavelength Range
405/450
Dye Type
Alexa Fluor™ Plus 405
Product Line
Alexa Fluor™ Plus
Shipping Condition
Qualified for Ambient or Wet Ice
Label Type
Alexa Fluor Dyes
Product Type
Phalloidin
SubCellular Localization
Actin

Contents & Storage

1 vial sufficient for 300 units/slides
Store at ≤-20°C after receiving
Desiccate, and protect from light
When stored as directed, product is stable for at least 1 year.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text