See more signal from immunofluorescence images with Prolong® Diamond. Learn more.

Alexa Fluor® 647 Goat Anti-Rabbit SFX Kit, highly cross-adsorbed

Catalog number: A31634

Invitrogen™

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Alexa Fluor® SFX Kits contain Image-iT® FX signal enhancer (Cat. no. I36933) plus one of sixteen different Alexa Fluor® dye-labeled secondary antibodies. These kits provide 400 µg (0.2 mL of 2 mg/mL) of either goat anti-mouse IgG or goat anti-rabbit IgG antibody (as a standard or highly cross-adsorbed preparation) conjugated to Alexa Fluor® 488, Alexa Fluor® 555, Alexa Fluor® 594, or Alexa Fluor® 647 dye, four of our most commonly used Alexa Fluor® dyes. The Alexa Fluor® 647 goat anti-rabbit IgG, highly cross-adsorbed, included with this kit is also available in a 1000 µg unit size (Cat. no. A21245).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Antibody Fragment: Whole Antibody
Host Species: Goat
Label or Dye: Alexa Fluor™ 647
Product Line: Alexa Fluor®, Molecular Probes®
Product Size: 1 kit
Reactivity: Rabbit
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products