Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
BioProbes™ Journal of Cell Biology Applications—Subscribe today »
Invitrogen™

Alexa Fluor™ 488 Streptavidin, 10 nm colloidal gold conjugate

Catalog number:  A32361

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Alexa Fluor® 488 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor® dye). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays. Alexa Fluor® dye streptavidin conjugates are supplied as 1 mg lyophilized product or in 0.5 mL volumes of a 2 mg/mL solution.

Important Features of Alexa Fluor® 488 Streptavidin Conjugates:
Alexa Fluor® 488 streptavidin conjugate has Ex/Em maxima of ~ (495/519)
Bright, photostable fluorescence
High solubility in aqueous solutions
Available in multiple colors
Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more

Properties of Alexa Fluor® Dyes
Alexa Fluor® dyes are organic fluorescent dyes developed for better performance in imaging and other labeling protocols and exhibit improved photostability and brightness and improved solubility in aqueous solutions. Available in a broad range of colors, these dyes are a good choice for most imaging applications.

Blocking Endogenous Biotin
Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Related Links:

Learn more about Avidin-Biotin Detection

Learn more about Alexa Fluor® Dyes

Find out about other Labeled Streptavidin Conjugates

Read Avidin and Streptavidin Conjugates-Section 7.6 in the Molecular Probes Handbook
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Alexa Fluor™ 488, Colloidal Gold
Concentration: 30 µg⁄ml
Product Line: Alexa Fluor™
Product Size: 0.5 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products