Invitrogen™

Alexa Fluor™ 488 Reactive Dye Decapack, for microarrays

Catalog number:  A32750

 Related applications: Genotyping & Genomic Profiling | Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Alexa Fluor® 488 reactive dye matches the imaging channels of most popular microarray scanners. The Alexa Fluor® reactive dye decapack is specially packaged succinimidyl ester dye for labeling of DNA or RNA samples that have been modified by incorporation of aminoallyl-dUTP (Cat. no. A21664) or aminohexylacrylamido-dUTP (Cat. no. A32761). Every packaging lot is rigorously tested for the ability to label aminoallyl-modified DNA to high and consistent levels.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Alexa Fluor™ 488
Labeling Method: Indirect Labeling
Product Line: Alexa Fluor™
Product Size: 1 set
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols