Alexa Fluor® 555-aha-dCTP

Catalog number: A32770

Molecular Probes™  Related applications: Genotyping & Genomic Profiling | Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Alexa Fluor® 555 dyes, used to label nucleotides, are compatible with commonly used microarray scanners, and provide greater signal correlation (R2) values than the spectrally similar Cy™3 and Cy™5 dye pair, improving the resolution of two-color microarray gene expression assays to 1.3-fold changes in expression. The exceptionally bright and photostable Alexa Fluor® dyes are also essentially insensitive tob pH and are highly water soluble.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration: 1 mM
Label or Dye: Alexa Fluor™ 555
Labeling Method: Direct Labeling
Modification: AHA (5-aminohexylacrylamido)
Product Line: Alexa Fluor®
Product Size: 50 µL
Shipping Condition: Wet Ice

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products