Consider upgrading to TrueCut Cas9 Protein v2 for higher editing efficiency.
Gibco™

StemFlex™ Medium

Catalog number:  A3349401

 Related applications: Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Gibco™ StemFlex™ Medium supports the robust expansion of feeder-free pluripotent stem cells (PSCs) and is optimized to support novel applications, including single-cell passaging, gene editing, and reprogramming. StemFlex Medium’s unique formulation offers the convenience of a flexible feeding schedule (including weekend-free options) and also the ability to choose the matrix and passaging reagent that best suits specific applications. StemFlex Medium enables the long term feeder-free culture of PSCs without karyotypic abnormalities and maintains the cells' ability to differentiate into all three germ layers.

Superior results
StemFlex Medium is formulated to deliver superior outcomes in today’s more-challenging applications that stress PSC cultures, such as reprogramming, single cell passaging, and gene editing. StemFlex Medium provides superior cell survival compared to the leading competitor (mTeSR1), allowing for greater success in the most relevant cell types.

Flexibility
StemFlex Medium allows for unprecedented 3-way usage flexibility for PSC culture conditions: the ability to select an optimal feeding schedule, including a weekend-free option; a choice of matrices, depending upon the needs of the workflow; and the option to select the passaging reagent for the level of cell dissociation required.

Exceptional value and performance
StemFlex Medium is provided in a convenient two-component kit (450 mL basal medium & 50 mL supplement), and when used with Geltrex™ LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix, provides a cost-effective, robust system for superior feeder-free culture of human PSCs. StemFlex Medium’s ability to enable weekend-free culture of PSCs provides unmatched value among the robust feeder-fee PSC media on the market today.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Stem Cells (Embryonic), Stem Cells (Human), Stem Cells (iPS - Induced Pluripotent Stem)
Product Line: StemFlex™
Product Size: 500 mL
Shipping Condition: Supplement ships on dry ice, basal can ship at RT

Contents & storage

1 x 450 mL Basal Medium, store at 2–8°C
1 x 50 mL Supplement, store at -20°C

Documents

Manuals & protocols

Recommended products