Amplex® ELISA Development Kit for Mouse IgG, with Amplex® UltraRed Reagent

Catalog number: A33851

Molecular Probes™  Related applications: Labeling Chemistry | Protein Assays and Analysis | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Amplex® ELISA Development Kit for Mouse IgG provides a comprehensive set of components for creating an absorbance or fluorescence-based ELISA using a mouse primary antibody. The assay is based on Amplex® UltraRed reagent, a fluorogenic or chromogenic for horseradish peroxidase (HRP) that reacts with H2O2 in a 1:1 stoichiometric ratio to produce Amplex®UltroxRed product, a brightly fluorescent and strongly absorbing reaction product (excitation/emission maxima ~568/581 nm). When used in fluorescence-based assay, Amplex® UltraRed reagent delivers better sensitivity and a broader assay range than colorimetric reagents.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 1 kit
Detection Method: Fluorescent
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols