ANTS (8-Aminonaphthalene-1,3,6-Trisulfonic Acid, Disodium Salt)

Catalog number:  A350

Invitrogen™

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The polyanionic dye ANTS is often used in combination with the cationic quencher DPX (X1525) for membrane fusion or permeability assays, including complement-mediated immune lysis. The unusually high Stokes shift of ANTS in water (>150 nm) separates this from much of the autofluorescence of biological samples. ANTS has also been employed as a neuronal tracer.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Green
For Use With (Equipment): Fluorescence Microscope
Sub-Cellular Localization: Cell Membranes & Lipids
Detection Method: Fluorescent
Excitation⁄Emission (nm): 350⁄520
Product Size: 1 g
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products