Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Invitrogen™

Acrylodan (6-Acryloyl-2-Dimethylaminonaphthalene)

Catalog number:  A433

 Related applications: Labeling Chemistry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The thiol reactive acrylodan (6-acryloyl-2-dimethylaminonaphthalene) generally reacts with thiols more slowly than iodoacetamides or maleimides, but does form very strong thioether bonds that are expected to remain stable under conditions required for complete amino acid analysis. The fluorescence emission peak and intensity of these adducts are particularly sensitive to conformational changes or ligand binding, making this dye one of the most useful thiol-reactive probe for protein structure studies. The environment-sensitive spectral shift of acrylodan conjugates may make this useful for distinguishing thiols that are located in membranes versus those exposed to aqueous solvation in cells.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 25 mg
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature and protect from light.

Documents

Manuals & protocols

Recommended products