Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

4-Acetamido-4'-Maleimidylstilbene-2,2'-Disulfonic Acid, Disodium Salt

Numéro de catalogue:  A485

Invitrogen™  Applications liées: Labeling Chemistry | Protein Labeling & Crosslinking

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

Our 4-acetamido-4'-maleimidylstilbene-2,2'- disulfonic acid is a thiol-reactive reagent that is water soluble, with high polarity and membrane impermeability. This polysulfonated dye is useful for determining whether thiol-containing proteins and polypeptide chains are exposed at the extracellular or cytoplasmic membrane surface.
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Chemical Reactivity: Thiol
Label or Dye: Stilbene
Product Size: 25 mg
Shipping Condition: Room Temperature

Contenu et stockage

Store in freezer (-5 to -30°C).

Documents

Manuels et protocoles

Produits recommandés