Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

2-Aminoacridone, Hydrochloride

Numéro de catalogue:  A6289

Invitrogen™  Applications liées: Labeling Chemistry

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

2-Aminoacridone is a small, amine-containing fluorophore that is used to label reducing sugars, oligosaccharides, and glycosphingolipids. Subpicomolar detection of the derivatized products has been reported by fluorophore-assisted carbohydrate electrophoresis (FACE). Other key applications for this dye include capillary electrophoresis (CE) and high-performance liquid chromatography (HPLC)
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Label or Dye: Acridone
Chemical Reactivity: Aldehyde, Ketone
Product Size: 25 mg
Shipping Condition: Room Temperature

Contenu et stockage

Store at room temperature and protect from light.

Documents

Manuels et protocoles

Produits recommandés