Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Avidin, NeutrAvidin®, Oregon Green® 488 conjugate

Catalog number: A6374

Invitrogen™

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Our Oregon Green® 488 conjugate of NeutrAvidin® biotin-binding protein - a form of avidin that has been processed to remove carbohydrate and to lower its isoelectric point - can substantially decrease background due to nonspecific binding. The method used to deglycosylate the avidin retains its specific binding. This bright, green-fluorescent reagent (absorption/emission maxima ~496/524 nm) is ideal for detecting biotinylated targets in fluorescence microscopy and flow cytometry applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Oregon Green® 488
Product Line: NeutrAvidin®, Oregon Green®
Product Size: 1 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products