Thermo Scientific™
HistoStar™ Embedding Workstation
HistoStar™ Embedding Workstation
 Related applications: Anatomical Pathology
Embed routine tissue samples into paraffin blocks for microtome sectioning comfortably and efficiently with the Thermo Scientific™ HistoStar™ Embedding Workstation. This workstation reduces fatigue and potential errors common when using other uncomfortable, hot or poorly illuminated embedding centers.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Brightly Illuminated and Clutter Free 
  • User-adjustable LED lighting uniformly illuminates the workspace without the clutter of awkward remote lamps, reducing fatigue and minimizing errors.
Cool and Contoured
  • Smooth and insulated user contact points provide a cool, comfortable workspace.
  • Constructed to eliminate pressure points and uncomfortable heat conditions.
High Capacity for High Efficiency
  • 5 liter paraffin capacity, coldplate area for 72 base molds, large heated workspace and specimen holding area enable users to manage heavy workflow in even the busiest laboratories.
Handle Any Sample Size with Ease
  • Workstation offers the first adjustable paraffin dispense paddle.
  • A simple adjustment moves paddle to the most ergonomic position for each user.
  • Embed even Super Mega Cassettes with ease.
Integrated ParaTrimmer
  • Only workstation with a heated wax trimmer built directly into the workspace.
  • Remove excess paraffin at the embedding station, eliminating the need for additional equipment or wax scraping at the microtome.
Intuitive User Interface
  • Large, easy-to-read touchscreen display allows quick access to temperature controls and other parameters.
  • Programmable asleep mode saves energy during workflow lapses.
Dependable Performance
  • Robust design minimizes moving parts, reducing wear and service requirements and ensuring dependable performance for years.
Documents
Manuals & protocols