Thermo Scientific™
SlideMate™ AS Slide Printer
 Related applications: Anatomical Pathology
Reduce specimen identification errors while enhancing lean workflow with the Thermo Scientific™ SlideMate™ AS On-Demand Slide Printer.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
The SlideMate AS is the next generation Thermo Scientific Slide Printer delivering a smaller footprint than previous models, making it even easier to fit next to the microtome. The SlideMate AS promotes single piece workflow though unique features designed specifically for on-demand printing.
  • Fully integrated color touch-screen and simple software are easy to use
  • Build-in barcode scanner reads cassette information and imports the data to the slide label
  • Connects easily to other computers or Laboratory Information Systems
  • Prints directly onto the slide, eliminating the need for hand writing or labels
Documents
Manuals & protocols