NucAway™ Spin Columns

Catalog number:  AM10070

Invitrogen™  Related applications: DNA Extraction | Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® NucAway™ Spin Columns are for the fast removal of unincorporated nucleotides and salts from probes after probe synthesis reactions. 30 columns are provided.
• Fast, efficient removal of unincorporated nucleotides and salts after probe synthesis
• RNase- and DNase-free
• Replaces more time- consuming purification methods To use, simply rehydrate the column, centrifuge to remove the interstitial fluid, add the probe to the top of the column, and centrifuge again to elute the probe.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: Spin Column
Product Line: Ambion™, NucAway™
Product Size: 30 each
Shipping Condition: Room Temperature

Contents & storage

Store tubes at room temperature.

Documents

Manuals & protocols

Recommended products