BrightStar®-Plus Positively Charged Nylon Membrane, 15 cm x 15 cm

Catalog number: AM10100

Ambion™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® BrightStar®-Plus Positively Charged Nylon Membrane(s) are strongly recommended to ensure consistent, reliable results with minimal background. Ideal for use with radiolabeled and nonisotopic probes for northern and other blot applications, five sheets measuring 15 x 15 cm are provided. This membrane exhibits the lowest nonspecific binding in nonisotopic detection applications when used with Ambion® blocking and wash solutions.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Membrane: Nylon Membrane
Product Size: 5 pieces
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols