Fluor-Coated TLC Plate (10 x 10 cm)

Catalog number: AM10110

Ambion™  Related applications: DNA Extraction | Nucleic Acid Gel Electrophoresis & Blotting | Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® Fluor-coated TLC plate UV shadowing is a quick and easy method for visualizing any nonisotopically labeled or unlabeled nucleic acid (DNA or RNA). Four 10 x 10 cm plates are provided. Simply place the plate underneath a gel and shine a short wavelength UV light down on the gel. The nucleic acid absorbs light at this wavelength and casts a shadow against the fluorescent background, allowing the full-length nucleic acid to be marked and excised from the gel. As little as 0.3 µg can be detected. Gel purification of full-length RNA transcripts away from smaller fragments for use in nuclease protection assays is recommended. UV shadowing is an ideal way to assess and gel purify nonisotopically labeled RNA probes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: TLC Plate
Plate Type: TLC Plate
Product Size: 4 plates
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols