KinaseMax™ 5' End-Labeling Kit

Catalog number: AM1520

Ambion™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting | Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Ambion® KinaseMax™ 5' End-Labeling Kit allows the efficient end-labeling of DNA or RNA to high-specific activity with T4 polynucleotide kinase and [gamma-32P] ATP, or quantitative phosphorylation of 5' ends using unlabeled ATP. Includes sufficient reagents for 30 reactions.

• Label oligonucleotide probes for nuclease protection assays and blot hybridizations
• Label primers for northerns and quantitative PCR
• Obtain up to 3-fold greater yields than with standard kinase buffers
• Reagents included for forward and dephosphorylation reactions

The kit includes a kinase reaction buffer that exceeds the performance of the standard forward reaction buffers recommended in "Molecular Cloning: A Laboratory Manual" (Sambrook et al., 1989) and "Current Protocols in Molecular Biology" (John Wiley & Sons, Inc. Ausubel, FM et al. editors, 1994). Crude 7000 Ci/mmol [gamma-32P]ATP is compatible with this kit. End-labeled probes are more stable than internally labeled probes and can routinely be used in assays for two to four weeks.

Rapid, Phenol-Free Phosphatase Removal Step
Inactivating the Calf Intestinal Phosphatase (CIP) after the dephosphorylation reaction is a time-consuming and cumbersome procedure that usually requires phenol extraction and ethanol precipitation typically resulting in sample loss. The KinaseMax™ Kit now includes a novel Phosphatase Removal Reagent for quick and complete removal of CIP. After the dephosphorylation step, the reagent is added to the reaction, incubated at room temperature for 2 minutes. The tube is then spun for a minute in the microfuge and the supernatant transferred to a new tube for the kinasing reaction.

Accessory Products:
The NucAway™ Spin Columns (SKU# AM10070) are available for rapid purification of end-labeled probes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: 32P (Phosphorous-32)
Product Size: 30 reactions
Final Product: Probes (Labeled RNA), Probes (Labeled DNA)
Labeling Method: Direct Labeling
Labeling Target: cDNA, PCR Products, DNA (General), Oligos, RNA (General)
Detection Method: Radioactive
Includes Label or Dye: No
Shipping Condition: Dry Ice

Contents & storage

T4 Polynucleotide Kinase, Calf Intestine Alkaline Phosphatase, 10X Kinase Buffer, Dephosphorylation Buffer, Phosphatase Removal Reagent, RNA Transcript Control, pUC19⁄Sau3 A Markers, ATP Solution, Gel Loading Buffer II should be stored at –20°C. Nuclease-free water may be stored at any temperature.

Documents

Manuals & protocols