Invitrogen™

MEGAscript™ RNAi Kit

Catalog number:  AM1626

 Related applications: RNAi

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Ambion® MEGAscript® RNAi Kit is for the synthesis of large mass amounts of dsRNA for RNAi experiments in nonmammalian systems. The MEGAscript RNAi Kit uses Ambion's patented high-yield transcription technology and includes reagents for transcription, nuclease digestion, and clean up. The kit includes sufficient reagents for 20 reactions.

Synthesize Large Amounts of dsRNA:
The ability to generate large amounts of clean, double-stranded RNA is essential for the success of RNAi experiments. The MEGAscript RNAi Kit allows RNAi researchers to synthesize 50 to 100 µg or more of double-stranded RNA in a single transcription reaction. The yield is typically 10–50 times the amount of RNA produced by conventional transcription reactions. DNA of either PCR or plasmid origin can be used as a template for the transcription reaction. The sense and antisense strands can be synthesized from a PCR-generated DNA template containing T7 RNA Polymerase promoters on both ends of the template, eliminating the need for a separate annealing step.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Control Type: Transcription Control
Product Line: Ambion™, MEGAscript®
Product Size: 20 reactions
Sample Type (Specific): Double-Stranded RNA (Long)
Shipping Condition: Dry Ice

Contents & storage

T7 Enzyme Mix, 10X T7 Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, 10X Binding Buffer, Elution Solution, RNase, DNase I, 10X Digestion Buffer, 2X Wash Solution, and Control Template are all stored at –20°C, Nuclease-free Water may be store at any temperature. Filter Cartridges and Collection Tubes are stored at room temperature.

Documents

Manuals & protocols

Recommended products