Invitrogen™

FirstChoice™ RLM-RACE Kit

Catalog number:  AM1700

 Related applications: Reverse Transcription | cDNA Libraries & Library Construction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The FirstChoice® RLM-RACE Kit is designed to amplify cDNA only from full-length, capped mRNA, usually producing a single band after PCR. This kit is a major improvement over the basic rapid amplification of cDNA ends (RACE) protocol. The RLM-RACE procedure selects only full-length mRNA—no rRNA, tRNA or degraded RNA—and facilitates the cloning of sequences from the 5' ends of messages.

Features of the FirstChoice® RLM-RACE Kit:

• From RNA to PCR product in less than a day
• Yields single, specific product from rare transcripts
• Selects 5' and/or 3' ends of true messages
• Efficient—all enzymatic reactions are optimized to ensure detection of even the rarest mRNA

RACE
Rapid amplification of cDNA ends (5'-RACE) is a polymerase chain reaction-based technique developed to facilitate the cloning of sequence from the 5'-ends of messages. The FirstChoice® RLM-RACE Kit is a major improvement to the basic RACE protocol.

Selects full-length mRNA—no rRNA, tRNA or degraded mRNA
In the FirstChoice® RLM-RACE procedure, full-length mRNAs are selected by treating total or poly(A) RNA with calf intestinal phosphatase (CIP) to remove the 5'-phosphate from all molecules which contain free 5'-phosphates (ribosomal RNA, fragmented mRNA, tRNA, and contaminating genomic DNA). Full-length mRNAs are unaffected. The RNA is then treated with tobacco acid pyrophosphatase (TAP) to remove the cap structure from the full-length mRNA leaving a 5'-monophosphate. A synthetic RNA adapter is ligated to the RNA population—only molecules containing a 5'-phosphate, the uncapped, full-length mRNAs, will accept the adapter. Random-primed, reverse transcription reactions and nested PCR are then performed to amplify the 5'-end of your specific transcript.

From RNA to PCR in less than a day
Each step of the FirstChoice® RLM-RACE procedure has been optimized, so you can complete all the enzymatic reactions and even start PCR in no more than 5 hours. The FirstChoice® RLM-RACE Kit contains reagents and enzymes to produce 5 RLM-RACE-ready cDNA preparations, in addition to primers and nested RACE adapter primers for 100 PCR reactions. Reverse transcription reagents are also included. Each kit contains control RNA and primers to test the kit's performance along with a detailed Instruction Manual. SuperTaq™ Thermostable Taq DNA Polymerase is available separately. For optimal amplification of fragments ≥1 kb, use SuperTaq-Plus. Also included are a 3' RACE Adapter and Primers for 3' RACE.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Final Product: cDNA
Final Product Size(s): 7 kb or less
Optimal Reaction Temperature: 42° C
Product Line: Ambion™, FirstChoice®
Product Size: 1 kit
Reaction Format: Separate Components
Reverse Transcriptase: M-MLV
Sample Type (General): RNA
Shipping Condition: Dry Ice

Contents & storage

The following components should be stored at –20°C:
  • Calf Intestine Alkaline Phosphatase
  • 10X CIP Buffer
  • Tobacco Acid Pyrophosphatase
  • 10X TAP Buffer, T4 RNA Ligase
  • 10X T4 RNA Ligase Buffer
  • 5' RACE Adapter
  • 3' RACE Adapter
  • 5' RACE Outer Primer
  • 5' RACE Inner Primer
  • 3' RACE Outer Primer
  • 3' RACE Inner Primer
  • Ammonium Acetate Stop Solution
  • M-MLV Reverse Transcriptase
  • dNTP Mix
  • 10X RT Buffer
  • 10X PCR Buffer
  • Random Decamers
  • RNase Inhibitor
  • Mouse Thymus RNA
  • 5' RACE Outer Control Primer
  • 5' RACE Inner Control Primer
  • 5' PCR Control Primer
  • 3' RACE Control Primer
Nuclease-free water may be stored at any temperature.

Documents

Manuals & protocols

Recommended products