Invitrogen™

Pre-miR™ hsa-miR-1 miRNA Precursor Postive Control

Catalog number:  AM17150

 Related applications: miRNA & Non-Coding RNA Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Pre-miR™ hsa-miR-1 miRNA Precursor is intended for use as a positive control in experiments utilizing Pre-miR miRNA Precursors. This precursor was chosen as a positive control because, upon delivery into cells, it effectively down regulates the expression of twinfilin-1, also known as PTK9, at the mRNA level (60-95% reduction. Effective delivery and activity of the Pre-miR hsa-miR-1 miRNA Precursor can be detected by real-time PCR using a TaqMan® Gene Expression Assay to PTK9 (Assay IDs: Hs00702289_s1 [human], Mm01598980_g1 [mouse]).

About Pre-miR miRNA Precursors

Pre-miR™ miRNA Precursor Molecules are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs. These ready-to-use miRNA mimics, which are similar to, but not identical to siRNAs, can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. They enable detailed study of miRNA biological effects via gain-of-function experiments (Figure 1). Positive and negative control Pre-miR miRNA Precursors (listed above) or the Pre-miR™ Precursor Starter Kit can be used to optimize transfection of Pre-miR miRNA Precursors.

Pre-miR miRNA Precursor Molecules have been carefully designed and modified to ensure that the correct strand, representing the desired mature miRNA, is taken up into the RNA-induced silencing-like complex responsible for miRNA activity (Figure 2). In contrast to miRNA expression vectors, these synthetic molecules can be used in dose response studies due to their introduction directly into the cell by transfection or electroporation. Thanks to their small size, they are easier to transfect than vectors, and can be delivered using conditions identical to those used for siRNAs.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Control Type: Gene Knockdown Positive Control
Label or Dye: Unconjugated
Product Line: Ambion™, Pre-miR™
Product Size: 5 nmol
RNAi Type: Pre-miR™ miRNA Precursor
Shipping Condition: Room Temperature

Contents & storage

Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.

Documents

Manuals & protocols