MagMAX™-96 AI/ND Viral RNA Isolation Kit

Catalog number: AM1835

Applied Biosystems™  Related applications: Automated Nucleic Acid Purification | Viral RNA⁄DNA Purification

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Applied Biosystems® This kit is for the isolation of viral RNA and DNA from diverse sample types (i.e., environmental samples and biofluids). Nucleic acids purified using this kit are ideal for PCR and RT-PCR applications. This is a first generation MagMAX™ Viral RNA Isolation Kit validated by the NVSL for use with AI/ND viruses. Perfect for working with low viral concentrations, the kit contains sufficient reagents for 4 x 96 reactions.

• Validated by the NVSL for use with AI/ND viruses
• Recover as few as 10 copies of viral RNA from 100–400 µL sample
• Typical viral RNA recovery of >50%
• Ideal when working with low viral concentrations
• Linear recovery down to 50 transcripts
• 96 samples can be processed in 30 minutes
• Easily integrated into robotic platforms

Benefits of MagMAX™ Magnetic Bead-Based Purification:
Magnetic beads offer many benefits compared to other technologies for isolating RNA. Beads bind RNA more efficiently than glass fiber filters, resulting in higher and more consistent RNA yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples. Since only a small volume of magnetic beads is needed for high-efficiency binding, the bound RNA can be eluted in just 20–50 µL of nuclease-free water, concentrating RNA from large, dilute samples.

Optimized for Isolation of Virus from Biofluids:
The Applied Biosystems® MagMAX™ Viral RNA Isolation Kit has been optimized for use with biological fluids and cell-free samples such as serum, plasma, swabs, and cell culture media. These samples often present unique challenges when purifying RNA, including low viral titers, large volumes to be processed, and sample complexity. Without the need for organic solvents or RNA precipitation, the MagMAX™ Viral RNA isolation kit eliminates the problems often encountered with other RNA isolation methods such filter clogging, large elution volumes, and inconsistent yields.

Get the Same Amount of RNA from 10-fold Less Sample
The sensitivity of viral RNA detection was compared using RNA isolated with Applied Biosystems® MagMAX™ protocol or the current National Veterinary Services Laboratories (NVSL) validated viral testing protocol. Even at one-tenth the sample input, viral RNA isolated using the MagMAX™ kit could be detected at equal or greater levels than RNA isolated using the NVSL protocol and recommended competitor's kit (see data). Downloadable protocols for the use of this kit with specific liquid handling systems are available at our Automation Resource.

Accessory Products:
Carrier RNA (SKU#4382878), Lysis/Binding Solution Concentrate (SKU#AM8500), Wash Solution 1 Concentrate (SKU#AM8504), and Wash Solution 2 Concentrate (SKU#AM8640) are components of the MagMAX™ Viral RNA Isolation Kits that may also be purchased separately. A 96-well magnetic ring stand is required to use this kit. The Magnetic Stand-96 (SKU#AM10027) and the 96-well Magnetic Ring Stand (SKU#AM10050) are designed for paramagnetic bead precipitation from standard 96 well U-bottom microplates. The Magnetic-Ring Stand pellets the magnetic beads in a circle shape, enabling easy removal of supernatant by a robot or multichannel pipettors. For additional processing plates, Matrix 96-Well U Bottom Polypropylene Plates (Thermo Scientific, Item# 4917) are recommended. The MagMAX™ and MagMAX™-96 Viral RNA Isolation Kits (SKU#AM1939 and SKU#AM1836) are the newest generation MagMAX™ kits for this application, but are not yet validated by the NVSL.
For Veterinary Use Only.

Specifications

Sample Type (General): Liquid Samples (e.g. Serum), Viral Samples
Sample Type (Specific): Any Swab Type, Cell-Free Fluids, Plasma, Serum
Final Product: Viral RNA
Downstream Application: Northern Blotting, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR)
For Use With (Equipment): KingFisher™ Flex Magnetic Particle Processor, MagMAX™ Express-96 Standard Magnetic Particle Processor
High Throughput Compatibility: Automated Protocols, High Throughput-Compatible, Not High Throughput-Compatible (Manual)
Isolation Technology: Magnetic Bead
Format: 96-well plate
Number of Reactions: 384 Reactions
Product Line: MagMAX™
Product Size: 384 preps
Purification Time: 1 hrs
Starting Material (Amount): Up to 50µl of sample
Yield (up to): 5 ng

Contents & storage

Carrier RNA is stored at –20°C; RNA Binding Beads are stored at 4°C. Wash Solution 2 Concentrate and Elution Buffer may be stored at either 4°C or room temperature. Lysis⁄Binding Solution Concentrate, Wash Solution 1 Concentrate, RNA Rebinding Concentrate, Bead Resuspension Solution, Processing Plates and Lids, and PBS Buffer are stored at room temperature. Nuclease-free water may be stored at any temperature.

Documents

Manuals & protocols