Poly(A)Purist™ MAG Kit with Manual

Catalog number: AM1922M

Ambion™  Related applications: RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® Poly(A)Purist™ MAG Kit (patent pending) uses magnetic technology for the selection of mRNA that typically requires only a single round of selection yet yields RNA that is more highly enriched and devoid of rRNA than other procedures. Each kit contains sufficient reagents for the isolation of mRNA from up to 8 mg of total RNA.

• Highest recovery of mRNA of any available kit
• High stringency for lowering residual rRNA content
• Single round of selection sufficient for most applications

The superior performance of Poly(A)Purist™ MAG Kit is clearly demonstrated by a comparison of the mRNA recovered using 4 other kits (see Figure). The kit uses batch binding of RNA to premeasured aliquots of oligo(dT) cellulose to circumvent the problems of slow flow rates and clogged columns often experienced during conventional, gravity-driven chromatography. The magnetic bead–based selection employed in the delivers increased processing speed, ease of handling, and scalability. The entire selection procedure can be completed in as little as 45 minutes (see Figure).

Accessory Products:
A magnetic stand is required for use this kit. They are available in several formats, including the Single Place Magnetic Stand (SKU #AM10026) and the 6 Tube Magnetic Stand (SKU #AM10055).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): RNA
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Downstream Application: In Vitro Translation, Microarray Analysis, Microinjection, Northern Blotting, Nuclease Protection Assays, Nucleic Acid Labeling, Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction
Final Product: mRNA
Isolation Technology: Magnetic Bead, Oligo dT Affinity
Product Line: Ambion™, Poly(A)Purist™
Product Size: 1 kit

Contents & storage

Store Nuclease–free Water, Microfuge Tubes and Microfuge tubes with Spin Columns at room temperature. Store 2X Binding Solution, Wash Solution 1, Wash Solution 2 and Oligo(dT) Cellulose at 4°C. Store RNA Storage Solution, 5M Ammonium Acetate and Glycogen(5mg⁄mL) at -20°C.

Documents

Manuals & protocols

Recommended products