Recover RNA and gDNA from the same FFPE section—see the new user bulletin for protocol.
New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

RecoverAll™ Total Nucleic Acid Isolation Kit for FFPE

Catalog number:  AM1975

 Related applications: RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The RecoverAll™ Total Nucleic Acid Isolation Kit for FFPE is for the extraction of total nucleic acid from formalin- or paraformalin-fixed, paraffin-embedded (FFPE) tissues. Sufficient reagents are included for 40 purifications from up to four 20 µm sections, or up to 35 mg of unsectioned, core samples each. Features of the RecoverAll™ Total Nucleic Acid Isolation Kit for FFPE:

• Optimized for isolation of total nucleic acids, including microRNAs, from FFPE tissue
• No overnight Proteinase K digestion required—deparaffinize in the morning and perform qRT-PCR in the afternoon
• Typical yields are >50% of those of unfixed tissue
• Recovered nucleic acids are suitable for next generation sequencing, real-time RT-PCR, PCR, mutation screening, and microarray analyses

Extraction of nucleic acids from difficult samples
Archived tissue samples contain valuable information of disease states, but it has traditionally been difficult to isolate nucleic acids from them of a quality suitable for molecular analysis. Standard preservation techniques use formalin that maintains tissue structure and prevents putrefaction, but which also traps nucleic acids and modifies them through protein-protein and protein-nucleic acid crosslinks. RNA (and to some extent DNA) is often so fragmented and chemically modified that it is incompatible with many molecular analysis techniques. RNA fragmentation in FFPE tissues cannot be reversed; however, the protease digestion conditions of the RecoverAll™ kit are designed to release a maximal amount (see figure) of trapped RNA fragments of all sizes, including microRNA, in a relatively short amount of time.

The RecoverAll™ Total Nucleic Acid Isolation Kit solution
The RecoverAll™ Total Nucleic Acid Isolation Kit procedure requires about 45 minutes of hands-on time and can be completed in typically less than 1 day for RNA. FFPE samples are deparaffinized using a series of xylene and ethanol washes. Next, they are subjected to an extensive protease digestion with an incubation time tailored for the recovery of either RNA or DNA. Nucleic acids are then purified using a rapid glass-filter method, and are eluted into either water or low-salt buffer.

Nucleic acids for almost any downstream application
As is the case with all FFPE tissue, sample fixation and storage typically cause nucleic acid fragmentation and modification. Therefore, downstream applications, such as microarray analysis, which require more pristine RNA than does qRT-PCR, may require modification for optimal results. Although DNA tends not to fragment as easily as RNA, it appears to be more reactive to the formalin and requires a longer (2-day) protease digestion time to release substantial amounts of DNA.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): FFPE & Fixed Samples
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Downstream Application: Next-Generation Sequencing, Northern Blotting, PCR, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Southern Blotting, cDNA Library Construction, microRNA Analysis
Final Product: Genomic DNA, Total RNA, micro RNA
Isolation Technology: Spin Column (Glass Fiber Filter)
Product Line: Ambion™, RecoverAll™
Product Size: 40 preps
Starting Material (Amount): Up to 35 mg of unsectioned core samples, Up to four 20 µm sections
Yield (up to): 24 µg

Contents & storage

• 16 ml Digestion Buffer (room temperature)
• 60 ml Wash 1 Concentrate (room temperature)
• 60 ml Wash 2/3 Concentrate (room temperature)
• 80 Collection Tubes (room temperature)
• 40 Filter Cartridges (room temperature)
• 19.2 ml Isolation Additive (room temperature)
• 5 ml Elution Solution (-20°C, 4°C, or room temperature)
• 160 µl Protease (-20°C)
• 240 µl 10X DNase Buffer (-20°C)
• 160 µl DNase (-20°C)
• 400 µl RNase A (-20°C)

This kit is shipped in two parts: one at room temperature and one at -20°C.

Documents

Manuals & protocols

Recommended products