Invitrogen™

GLOBINclear™ Kit, human, for globin mRNA depletion

Catalog number:  AM1980

 Related applications: RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The GLOBINclear™-Human Kit uses a novel, non-enzymatic technology that rapidly depletes >95% of the alpha and beta globin mRNA from total RNA preparations derived from whole blood. Advantages of the GLOBINclear™-Human Kit:

• Removes >95% of unwanted globin mRNA from whole blood total RNA
• Detects up to 50% more genes—analyze previously undetected genes
• Achieves greater representation of the genome, low 3'/5' ratios
• Avoids RNase H treatments that can degrade mRNA and alter expression profiles

Using whole blood total RNA samples
It is widely known that expression array data generated from whole blood total RNA samples have reduced detection sensitivity compared to data from fractionated blood samples. Examination of whole blood total RNA reveals a dominant band in the 600–700 base region. This band represents globin mRNA, which is expressed at high levels in red blood cells (RBCs) and reticulocytes (RBC precursors). Up to 70% of the mRNA (by mass) in whole blood total RNA are globin transcripts. Array analysis using total RNA containing a high percentage of globin mRNA has been shown to decrease present calls, decrease call concordance, and increase signal variation.

Using the GLOBINclear Kit
The GLOBINclear™ kit uses a novel hybridization technology that takes advantage of the strength of biotin/streptavidin binding, the specificity of nucleic acid hybridization, and the convenience of magnetic separations. There are no harsh RNase H enzymatic treatments that may compromise the expression profile by degrading mRNAs other than globin mRNA. The resulting mRNA is a superior template for synthesizing labeled cDNA for array analysis and is ideal for quantitative RT-PCR. The enrichment delivers a significant increase in Percent Present calls, decreased sample-to-sample variability, and low 3'/5' ratios.

Accessory product
The kit requires the use of a magnetic stand. The DynaMag-2 Magnetic Stand (Cat. No. 12321D) is recommended.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): Blood, RNA
High Throughput Compatibility: High Throughput-Compatible
Downstream Application: Microarray Analysis, Northern Blotting, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction
Final Product: Total RNA (Globin mRNA-Depleted)
Format: 6 microtubes
Isolation Technology: Magnetic Bead
Number of Reactions: 20 Reactions
Product Line: Ambion™, GLOBINclear™
Product Size: 20 preps
Purification Time: 1.5 hrs
Sample Type (Specific): Total RNA (Whole Blood)
Scale: Mini
Starting Material (Amount): 1 - 10 µg of human whole blood total RNA
Yield (up to): 5 µg

Contents & storage

• 20 µl Capture Oligo Mix (-20°C)
• 600 µl Streptavidin Magnetic Beads (4°C—do not freeze)
• 1.5 ml 2X Hybridization Buffer (room temperature)
• 1.5 ml Streptavidin Bead Buffer (room temperature)
• 1.75 Nuclease-free Water (-20°C, 4°C, or room temperature)
• 200 µl RNA Binding Beads (4°C—do not freeze)
• 5 ml RNA Wash Solution Concentrate (4°C or room temperature)
• 1 ml Elution Buffer (4°C or room temperature)
• 80 µl RNA Bead Buffer (room temperature)
• 4 ml RNA Binding Buffer Concentrate (room temperature)
• 100 1.5-ml Non-stick Tubes (room temperature)

All components shipped on wet ice except for Capture Oligo Mix (dry ice).

Documents

Manuals & protocols