Invitrogen™

Poly(A) Polymerase (cloned) 2 U/μL

Catalog number:  AM2030

 Related applications: Nucleic Acid Labeling & Oligo Synthesis | Reverse Transcription

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® Poly(A) Polymerase catalyzes the addition of adenosine to the 3' end of RNA in a sequence-independent fashion. The enzyme can be used to append radiolabeled ATP to the 3' end of RNA molecules to generate labeled RNAs. One tube containing 80 U (2 U/ µL), 5X Poly(A) Polymerase Buffer, and 25 mM MnCl2 are provided. The enzyme can be used in conjunction with terminal transferase, reverse transcriptase, and thermostable polymerase to clone RNAs of unknown sequence.

Unit Definition:
One unit is the amount of enzyme required to incorporate 1 nmol of AMP into acid-insoluble material in 10 min at 37°C.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: Poly(A) RNA Polymerases
Product Line: Ambion™
Product Size: 80 units
Shipping Condition: Dry Ice

Contents & storage

Poly(A) Polymerase, 5X Poly(A) Polymerase Buffer, and 25 mM MnCl2 should be stored at –20°C.

Documents

Manuals & protocols

Recommended products