SuperTaq™ Polymerase (Cloned) 5 U/μl

Catalog number: AM2052

Applied Biosystems™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This high-purity Ambion® Taq Polymerase is isolated from an E. coli strain that over-expresses this gene. Its heat stability allows DNA polymerization in the 5' to 3' direction during repeated heat denaturation and reannealing steps in the presence of dNTPs, a DNA template, and complementary primers. It is supplied in one tube containing 250 U at a concentration of 5 U/ µL. It can be used in PCR and RT-PCR reactions.

Unit Definition:
One unit incorporates 10 nmol of deoxynucleotides into acid insoluble material in 30 min at 74°C. SuperTaq™ is a trademark of, and is manufactured by Enzyme Technologies Ltd. and sold under licensing arrangements with F. Hoffman-La Roche, Ltd., Roche Molecular Systems, Inc. and the Perkin-Elmer Corporation. We are a distributor of Enzyme Technologies, Ltd., except in the following countries: Austria, Benelux, Denmark, Greece, Sweden, Switzerland, Taiwan, and the United Kingdom.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: SuperTaq™ Polymerase
Hot Start: No
Fidelity (vs. Taq): 1 X
GC-Rich PCR Performance: Low
Reaction Format: Separate Components
Product Overhang: Mixed
Reaction Speed: Standard
Final Product Size(s): 2.5 kb or less
Product Size: 250 units
Exonuclease Activity: None
Shipping Condition: Dry Ice

Contents & storage

dNTPs, 10X Reaction Buffer + MgCl2, 10X Reaction Buffer - MgCl2, and MgCl2 are stored at –20°C.

Documents

Manuals & protocols