T7 RNA Polymerase (cloned) 20 U/μL

Catalog number: AM2082

Invitrogen™  Related applications: In Vitro Transcription | Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This cloned high-purity Ambion® RNA polymerase has a high specificity for its promoter. The enzyme will transcribe large amounts of RNA from DNA sequences downstream of the promoter and show no cross talk between promoters. One tube containing 5000 U at a concentration of 20 U/ µL is provided. Its high purity makes them ideal for synthesizing high specific activity RNA hybridization probes, biologically active mRNA, and antisense RNA. The RNA Polymerase is rigorously tested for contaminating nonspecific endonuclease, exonuclease, protease, and RNase activity. It is also tested for functionality with the MAXIscript® Kit at 20 U per reaction.

Unit Definition:
One unit of RNA Polymerase will catalyze the incorporation of 1 nmol of nucleoside triphosphate into acid-insoluble material in 60 min at 37°C.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Promoter: T7
Polymerase: T7 RNA Polymerase
Product Size: 5,000 units
Shipping Condition: Dry Ice

Contents & storage

Store at –20°C.

Documents

Manuals & protocols