Invitrogen™

Ambion™ RNase A, affinity purified, 1 mg/mL

Catalog number:  AM2270

 Related applications: DNA Extraction | Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® RNase A is an endonuclease that specifically cleaves 3' of U and C residues. Affinity Purified RNase A is intended for critical applications when the absence of DNase and other nonspecific nuclease activities is essential. Supplied in one tube containing 200 µg (1 mg/mL). Note: This preparation contains RNase B, a carbohydrate isoform of RNase A. RNase A is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Enzyme: RNase
Product Line: Ambion™
Product Size: 200 µg
Shipping Condition: Dry Ice

Contents & storage

Store at –20°C.

Documents

Manuals & protocols

Recommended products