Invitrogen™

Ribonuclease H, from E. coli (cloned) 10 U/μL

Catalog number:  AM2292

 Related applications: Real Time PCR (qPCR) | Reverse Transcription

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ambion® Ribonuclease H (RNase H) is isolated from an E. coli strain that over-expresses the gene. RNase H specifically degrades the RNA in RNA:DNA hybrids to produce 3' -hydroxyl and 5' -phosphate terminated products. It is supplied in one tube containing 200 U (10 U/ µL). The enzyme will not degrade DNA or unhybridized RNA. RNase H is an integral part of most RNA amplification and NASBA protocols. It can also be used to degrade specific RNAs when the complementary DNA oligo is hybridized, such as poly(A) tail removal from mRNA hybridized to oligo(dT). Ribonuclease H is rigorously tested for contaminating nonspecific endonuclease, exonuclease, RNase, and protease activity.

Unit Definition:
One unit of Ribonuclease H is the amount of enzyme required to increase fluorescence 1.5 RFUs per sec at 37°C using 20 pmol of RNaseAlert® probe coupled to 1,000 pmol of a complementary oligonucleotide as substrate.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Enzyme: RNase
Product Line: Ambion™
Product Size: 200 units
Shipping Condition: Dry Ice

Contents & storage

Store at –20°C

Documents

Manuals & protocols

Recommended products